Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can bind both GDIs for the close sequences.
Immunogen sequence: MNEEYDVIV LGTGLTECIL SGIMSVNGKK VLHMDRNPYY GGESASITPL EDLYKRFKIP GSPPESMGRG RDWNVDLIPK FLMANGQLVK MLLYTEVTRY LDFKVTEGSF VYKGGKIYKV PSTEAEALAS SLMGLFEKRR FRKFLVYVAN FDEKDPRTFE GIDPKKTTMR DVYKKFDLGQ DVIDFTGHAL ALYRTDDYLD QPCYETINRI KLYSESLARY GKSPYLYPLY GLG (1-232 aa encoded by BC005145)
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 46e; GDI beta; GDI-2; GDI-3; guanosine diphosphate (GDP) dissociation inhibitor 3; Guanosine diphosphate dissociation inhibitor 2; Rab GDI beta; Rab GDP dissociation inhibitor beta; RAB GDP dissociation inhibitor beta-2; rab GDP-dissociation inhibitor, beta
Gene Aliases: GDI-B; GDI2; Gdi3; GDIB; HEL-S-46e; RABGDIB
UniProt ID: (Human) P50395, (Mouse) Q61598
Entrez Gene ID: (Human) 2665, (Mouse) 14569
Molecular Function: G-protein modulator protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support