Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPRSFLVKS KKAHSYHQPR SPGPDYSLRL ENVPAPSRAD STSNAGGAKA EPRDRLSPES QLTEAPDRAS ASPDSCEGSV CERSSEFEDF WRPPSPSASP ASEKSMCPSL DEAQPFPLPF KPYSWSGLAG SDLRHLVQSY RPCGALERGA GLGLFCEPAP EPGHPAALYG PKRAAGGAGA GAPGSCSAGA GATAGPGLGL YGDFGSAAAG LYERPTAAAG LLYPERGHGL HADKGAGVKV ESELLCTRLL LGGGSYKCIK CSKVFSTPHG LEVHVRRSHS GTRPFACEMC GKTFGHAVSL EQHKAVHSQE RSFDCKICGK SFKRSSTLST HLLIHSDTRP YPCQYCGKRF HQKSDMKKHT FIHTGEKPHK CQVCGKAFSQ SSNLITHSRK HTGFKPFGCD LCGKGFQRKV DLRRHRETQH GLK (1-422 aa encoded by BC032751)
GFI1 is a nuclear zinc finger (ZF) transcriptional repressor that plays an important role in hematopoiesis and inner ear development, and has been implicated in lymphomagenesis. The Gfi1 protein expression is regulated by the ubiquitin-proteasome system. It is reported that ubiquitin ligase Triad1 interacts with the DNA-binding domain of Gfi-1. Gfi-1 represses transcription by directly binding to the consensus DNA sequence in the promoters of its target genes. Deficiency of Gfi-1 causes neutropenia and an accumulation of granulomonocytic precursor cells that is reminiscent of a myelodysplastic syndrome in mice.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ94509; growth factor independence-1; growth factor independent 1 transcription repressor; Growth factor independent protein 1; Growth factor independent-1; Zinc finger protein 163; Zinc finger protein Gfi-1
Gene Aliases: AW495828; GFI-1; GFI1; GFI1A; Pal-1; Pal1; SCN2; ZNF163
UniProt ID: (Human) Q99684, (Rat) Q07120, (Mouse) P70338
Entrez Gene ID: (Human) 2672, (Rat) 24388, (Mouse) 14581
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support