Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SYQGELKKGD TIYNTRTRKK VRLQRLARMH ADMMEDVEEV YAGDICALFG IDCASGDTFT DKANSGLSME SIHVPDPVIS I
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-Gmt; Elongation factor G 1, mitochondrial; Elongation factor G, mitochondrial; Elongation factor G1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM1; hEFG1; mEF-G 1; mitochondrial elongation factor G; mitochondrial elongation factor G1
Gene Aliases: COXPD1; EFG; EFG1; EFGM; EGF1; GFM; GFM1; hEFG1
UniProt ID: (Human) Q96RP9
Entrez Gene ID: (Human) 85476
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support