Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-6-phosphate amidotransferase 1; GFAT; GFAT 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine fructose aminotransferase; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine-fructose-6-phosphate aminotransferase 1; glutamine:fructose 6 phosphate amidotransferase 1; Glutamine:fructose-6-phosphate amidotransferase 1; Hexosephosphate aminotransferase 1
Gene Aliases: 2810423A18Rik; AI324119; AI449986; CMS12; CMSTA1; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; GFPT1; GFPT1L; MSLG
UniProt ID: (Human) Q06210, (Rat) P82808, (Mouse) P47856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support