Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IFEQPESVF NTMRGRVNFE TNTVLLGGLK DHLKEIRRCR RLIVIGCGTS YHAAVATRQV LEELTELPVM VELASDFLDR NTPVFRDDVC FFISQSGETA DTLLALRYCK DRGALTVGVT NTVGSSISRE TDCGVHINAG PEVGVASTKA YTSQFISLVM FGLMMSEDRI SLQNRRQEII RGLRSLPELI KEVLSLEEKI HDLALELYTQ RSLLVMGRGY NYATCLEGAL KIKEITYMHS EGILAGELKH GPLALIDKQM PVIMVIMKDP CFAKCQNALQ QVTARQGRPI ILCSKDDTES SKFAYKTIEL PHTVDCLQGI LSVIPLQLLS FHLAVLRGYD VDFPRNLAKS VTVE (330-682 aa encoded by B C000012)
GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-6-phosphate amidotransferase 2; GFAT 2; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 2; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine: fructose-6-phosphate aminotransferase 2; glutamine:fructose 6 phosphate amidotransferase 2; Glutamine:fructose-6-phosphate amidotransferase 2; Hexosephosphate aminotransferase 2
Gene Aliases: GFAT; GFAT 2; GFAT2; GFPT2
UniProt ID: (Human) O94808
Entrez Gene ID: (Human) 9945
Molecular Function:
transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support