Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MCGIFAYMNY RVPRTRKEIF ETLIKGLQRL EYRGYDSAGV AIDGNNHEVK ERHIQLVKKR GKVKALDEEL YKQDSMDLKV EFETHFGIAH TRWATHGVPS AVNSHPQRSD KGNEFVVIHN GIITNYKDLR KFLESKGYEF ESETDTETIA KLIKYVFDNR ETEDITFSTL VERVIQQLEG AFALVFKSVH YPGEAVATRR GSPLLIGVRS KYKLSTEQIP ILYRTCTLEN VKNICKTRMK RLDSSACLHA VGDKAVEFFF ASDASAIIEH TNRVIFLEDD DIAAVADGKL SIHRVKRSAS DDPSRAIQTL QMELQQIMKG NFSAFMQKEI FEQPESVFNT MRGRVNFETN TVLLGGLKDH LKEIRRCRRL IVIGCGTSYH AAVATRQVLE ELTELPVMVE LASDFLDRNT PVFRDDVCFF ISQSGETADT LLALRYCKDR GALTVGVTNT VGSSISRETD CGVHINAGPE VGVASTKAYT SQFISLVMFG LMMSEDRISL QNRRQEIIRG LRSLPELIKE VLSLEEKIHD LALELYTQRS LLVMGRGYNY ATCLEGALKI KEITYMHSEG ILAGELKHGP LALIDKQMPV IMVIMKDPCF AKCQNALQQV TARQGRPIIL CSKDDTESSK FAYKTIELPH TVDCLQGILS VIPLQLLSFH LAVLRGYDVD FPRNLAKSVT VE
GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-6-phosphate amidotransferase 2; GFAT 2; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 2; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine: fructose-6-phosphate aminotransferase 2; glutamine:fructose 6 phosphate amidotransferase 2; Glutamine:fructose-6-phosphate amidotransferase 2; Hexosephosphate aminotransferase 2
Gene Aliases: GFAT; GFAT 2; GFAT2; GFPT2
UniProt ID: (Human) O94808
Entrez Gene ID: (Human) 9945
Molecular Function:
transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support