Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAATAVAAAV AGTESAQGPP GPAASLELWL NKATDPSMSE QDWSAIQNFC EQVNTDPNGP THAPWLLAHK IQSPQEKEAL YALTVLEMCM NHCGEKFHSE VAKFRFLNEL IKVLSPKYLG SWATGKVKGR VIEILFSWTV WFPEDIKIRD AYQMLKKQGI IKQDPKLPVD KILPPPSPWP KSSIFDADEE KSKLLTRLLK SNHPEDLQAA NRLIKNLVKE EQEKSEKVSK RVSAVEEVRS HVKVLQEMLS MYRRPGQAPP DQEALQVVYE RCEKLRPTLF RLASDTTDDD DALAEILQAN DLLTQGVLLY KQVMEGRVTF GNRVTSSLGD IPVSRVFQNP AGCMKTCPLI DLEVDNGPAQ MGTVVPSLLH QDLAALGISD APVTGMVSGQ NCCEEKRNPS SSTLPGGGVQ NPSADRNLLD LLSPQPAPCP LNYVSQKSVP KEVPPGTKSS PGWSWEAGPL APSPSSQNTP LAQVFVPLES VKPSSLPPLI VYDRNGFRIL LHFSQTGAPG HPEVQVLLLT MMSTAPQPVW DIMFQVAVPK SMRVKLQPAS SSKLPAFSPL MPPAVISQML LLDNPHKEPI RLRYKLTFNQ GGQPFSEVGE VKDFPDLAVL GAA
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; Golgi-localized, gamma ear-containing, ARF-binding protein 2; Vear; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein
Gene Aliases: GGA2; KIAA1080; VEAR
UniProt ID: (Human) Q9UJY4
Entrez Gene ID: (Human) 23062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support