Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LSRPHGDTA KKPIIGILMQ KCRNKVMKNY GRYYIAASYV KYLESAGARV VPVRLDLTEK DYEILFKSIN GILFPGGSVD LRRSDYAKVA KIFYNLSIQS FDDGDYFPVW GTCLGFEELS LLISGECLLT ATDTVDVAMP LNFTGGQLHS RMFQNFPTEL LLSLAVEPLT ANFHKWSLSV KNFTMNEKLK KFFNVLTTNT DGKIEFISTM EGYKYPVYGV QWHPEKAPYE WKNLDGISHA PNAVKTAFYL AEFFVNEARK NNHHFKSESE EEKALIYQFS PIYTGNISSF QQCYIFD (23-318 aa encoded by BC025025)
Gamma-Glutamyl hydrolase (GGH) is a homodimeric protein that catalyzes the cleavage of the gamma-glutamyl chain of folylpoly-gamma-glutamyl substrates and is a central enzyme in folyl and antifolyl poly-gamma-glutamate metabolism. The activities of the baculovirus-expressed enzyme and the enzyme from HL-60 cells were similar on methotrexate polyglutamates. GGH is expressed in a variety of human tumors and correlates with poor prognosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Conjugase; FGPH; Folylpolyglutamate hydrolase; Gamma-Glu-X carboxypeptidase; Gamma-glutamyl hydrolase; gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase); GH
Gene Aliases: gamma-GH; GGH; GH
UniProt ID: (Human) Q92820, (Mouse) Q9Z0L8, (Rat) Q62867
Entrez Gene ID: (Human) 8836, (Mouse) 14590, (Rat) 25455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support