Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody is predicted to react with chicken, mouse and rat based on sequence homology.
May be involved in spermatogenesis. Its mouse homolog is induced by dioxin and may play a role in spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3HC4-type zinc finger protein; Gametogenetin-binding protein 2; laryngeal carcinoma related gene 1; Laryngeal carcinoma-related protein 1; Protein ZNF403; zinc finger protein 403
Gene Aliases: DIF-3; DIF3; GGNBP2; LCRG1; LZK1; ZFP403; ZNF403
UniProt ID: (Human) Q9H3C7
Entrez Gene ID: (Human) 79893
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support