Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Marmoset, Panda, Dog (94%) Zebu, Bovine (89%) Rabbit, Pig (83%).
Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GHRH receptor; Ghrh-r; GHRHRpsv; GRF receptor; Growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor
Gene Aliases: GHRFR; GHRHR; GRFR; IGHD1B
UniProt ID: (Human) Q02643
Entrez Gene ID: (Human) 2692
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support