Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: WGSVEEQRQQ QAELLAVIER LGREREGSFH SNDLFLDAQL LQRTGAGACQ EDYRQYQAKV EWQVEKHKQE LRENESNWAY K
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase IMAP family member 5; GTPase of the immunity-associated protein 5; IAN-4; IAN-5; immune associated nucleotide 4 like 1; Immune-associated nucleotide-binding protein 5; immunity associated protein 3; Immunity-associated nucleotide 4 protein; Immunity-associated nucleotide 4-like 1 protein; Immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5; lymphopenia
Gene Aliases: D630024P16; E230026N22Rik; GIMAP5; HIMAP3; IAN-5; IAN4; IAN4L1; IAN5; Iddm1; IMAP3; IROD; Lyp
UniProt ID: (Human) Q96F15, (Rat) Q8K3L6, (Mouse) Q8BWF2
Entrez Gene ID: (Human) 55340, (Rat) 246774, (Mouse) 317757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support