Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: MCF7, 293T, HeLa, THP-1, Mouse thymus, Rat fat
Immunogen sequence: LEKEKTRPEG EPSSLSPEEL AFAREFMANT ESYLKNVALK HMPPNLQKVD LFRAVPKPDL DSYVFLRVRE RQENILVEPD TDEQRDYVID LEKGSQHLIR
The yeast heterotetrameric GINS complex is made up of Sld5, Psf1 (GINS1), Psf2 (GINS2), and Psf3 (GINS3). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA replication complex GINS protein SLD5; GINS complex subunit 4; GINS complex subunit 4 (Sld5 homolog); SLD5; SLD5 homolog
Gene Aliases: 2810037C03Rik; 4933405K01Rik; GINS4; RGD1307397; SLD5
UniProt ID: (Human) Q9BRT9, (Mouse) Q99LZ3, (Rat) Q499W2
Entrez Gene ID: (Human) 84296, (Mouse) 109145, (Rat) 290842
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support