Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • GIPC1 Antibodies

            Proteintech

            GIPC1 Polyclonal Antibody

            9 References
            View all (14) GIPC1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite GIPC1 Polyclonal Antibody

            • Antibody Testing Data (8)
            Application
            GIPC1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 8

            GIPC1 Antibody (14822-1-AP) in ICC/IF

            Immunofluorescent analysis of HepG2 cells using 14822-1-AP (GIPC1 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            GIPC1 Antibody in Immunocytochemistry (ICC/IF)
            GIPC1 Antibody in Immunocytochemistry (ICC/IF)
            GIPC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            GIPC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            GIPC1 Antibody in Western Blot (WB)
            GIPC1 Antibody in Western Blot (WB)
            GIPC1 Antibody in Western Blot (WB)
            GIPC1 Antibody in Immunoprecipitation (IP)

            Product Details

            14822-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:600-1:4000
            View 6 publications 6 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 2 publications 2 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            GIPC1 Fusion Protein Ag6597 (1-333 aa encoded by BC000410)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.41 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MPLGLGRRK KAPPLVENEE AEPGRGGLGV GEPGPLGGGG SGGPQMGLPP PPPALRPRLV FHTQLAHGSP TGRIEGFTNV KELYGKIAEA FRLPTAEVMF CTLNTHKVDM DKLLGGQIGL EDFIFAHVKG QRKEVEVFKS EDALGLTITD NGAGYAFIKR IKEGSVIDHI HLISVGDMIE AINGQSLLGC RHYEVARLLK ELPRGRTFTL KLTEPRKAFD MISQRSAGGR PGSGPQLGTG RGTLRLRSRG PATVEDLPSA FEEKAIEKVD DLLESYMGIR DTELAATMVE LGKDKRNPDE LAEALDERLG DFAFPDEFVF DVWGAIGDAK VGRY (1-333 aa encoded by BC000410)

            Target Information

            GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008).

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: GAIP C-terminus-interacting protein; GIPC PDZ domain containing family, member 1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; IGF-1 receptor interacting protein 1; neurophilin1-IP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; RGS19-interacting protein 1; semaF cytoplasmic domain associated protein; SemaF cytoplasmic domain-associated protein 1; SEMCAP-1; Synectin; Tax interacting protein, clone 2; Tax interaction protein 2; TIP-2

            View more View less

            Gene Aliases: C19orf3; GIPC; GIPC1; GLUT1CBP; Glut1CIP; Hs.6454; IIP-1; NIP; Rgs19; RGS19IP1; SEMCAP; Semcap1; SYNECTIIN; SYNECTIN; TaxIP2; TIP-2

            View more View less

            UniProt ID: (Human) O14908, (Rat) Q9Z254, (Mouse) Q9Z0G0

            View more View less

            Entrez Gene ID: (Human) 10755, (Rat) 83823, (Mouse) 67903

            View more View less

            Function(s)

            actin binding receptor binding protein binding myosin binding PDZ domain binding protein homodimerization activity cadherin binding involved in cell-cell adhesion GTPase activator activity scaffold/adaptor protein

            Process(es)

            protein targeting G-protein coupled receptor signaling pathway chemical synaptic transmission glutamate secretion positive regulation of transforming growth factor beta receptor signaling pathway regulation of protein stability negative regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of cytokinesis endothelial cell migration regulation of synaptic plasticity cell-cell adhesion positive regulation of GTPase activity
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            • Fair Trade
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Korea flag icon
            Korea

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Thermo Fisher Scientific Korea Ltd.
            Representative : Soojin Seok
            Company Registration No. : 117-81-46910

             

            Thermo Fisher Scientific Solutions LLC
            Representative : Soojin Seok
            Company Registration No. : 114-86-04783

             

            Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0