Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
GTPase-activating protein for the ADP ribosylation factor family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARF GAP GIT2; ARF GTPase activating protein 2; ARF GTPase-activating protein GIT2; CAT-2; cool associated tyrosine phosphorylated-2; cool-associated, tyrosine phosphorylated protein 2; Cool-interacting tyrosine-phosphorylated protein 2; G protein-coupled receptor kinase interacting ArfGAP 2; G protein-coupled receptor kinase-interactor 2; GRK-interacting protein 2; Paxillin kinase linker
Gene Aliases: 1500036H07Rik; 5830420E16Rik; 6430510B20Rik; 9630056M03Rik; B230104M05Rik; CAT-2; CAT2; GIT2; KIAA0148; mKIAA0148; PKL
UniProt ID: (Human) Q14161, (Rat) Q66H91, (Mouse) Q9JLQ2
Entrez Gene ID: (Human) 9815, (Rat) 304546, (Mouse) 26431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support