Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAPKTAAVG PLVGAVVQGT NSTRFLVFNS KTAELLSHHK VELTQEFPKE GWVEQDPKEI LQSVYECIAR TCEKLDELNI DISNIKAVGV SNQRETTVIW DKLTGEPLYN AVVWLDLRTQ TTVEDLSKKI PGNSNFVKSK TGLPLSTYFS AVKLRWMLDN VRNVQKAVEE GRALFGTIDS WLIWSLTGGV NGGVHCTDVT NASRTMLFNI HSLEWDKELC DFFEIPMDLL PNVFSSSEIY GLIKTGALEG VPISGCLGDQ CAALVGQMCF QEGQAKNTYG TGCFLLCNTG RKCVFSEHGL LTTVAYKLGR EKPAYYALEG SVAIAGAVIR WLRDNLGIIE TSGDIERLAK EVGTSYGCYF VPAFSGLYAP YWEPSARGIL CGLTQFTNKC HIAFAALEAV CFQTREILEA MNRDCGIPLR HLQVDGGMTN NKVLMQLQAD ILHIPVIKPF MPETTALGAA MAAGAAEGVS VWSLEPQALS VLRMERFEPQ IQATESEIRY ATWKKAVMKS MGWVTSQSPE GGDPSIFSSL PLGFFIVSSM VMLIGARYIS GVP
GK2 (glycerol kinase 2), also known as GKP2 or GKTA, is a 553 amino acid protein that belongs to the FGGY kinase family and is involved in the pathway of glycerol degradation. Localized to the outer membrane of the mitochondrion and expressed at high levels in testis, GK2 functions to catalyze the ATPdependent conversion of glycerol to glycerol 3-phosphate. Via its catalytic activity, GK2 plays an essential role in the regulation of glycerol uptake and metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP:glycerol 3-phosphotransferase 2; GK 2; glycerokinase 2; Glycerol kinase 2; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; Glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m
Gene Aliases: GK2; GKP2; GKTA
UniProt ID: (Human) Q14410
Entrez Gene ID: (Human) 2712
Molecular Function:
carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support