Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LGVFLAPALA NYNINVNDDN NNAGSGQQSV SVNNEHNVAN VDNNNGWDSW NSIWDYGNGF AATRLFQKKT CIVHKMNKEV MPSIQSLDAL VKEKKLQGKG PGGPPPKGLM YSVNPNKVDD LSKFGKNIAN MCRGIPTYMA EEMQEASLFF YSGTCYTTSV L; Positive Samples: Mouse stomach
Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predominantly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18 kDa antrum mucosa protein; AMP-18; foveolin; Gastrokine-1; Protein CA11 homolog
Gene Aliases: 2200002K21Rik; AMP-18; Amp18; BRICD1; Ca11; Fov; Gkn1
UniProt ID: (Mouse) Q9CR36
Entrez Gene ID: (Mouse) 66283
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support