Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is 293 cell lysate.
PA5-20661 can be used with blocking peptide PEP-0781.
The proper expression of gene products in eukaryotic cells relies on efficient transport of mRNA molecules out of the nucleus. Gle1 is an essential mRNA export factor in both human and yeast cells. It associates with the nuclear pore complex (NPC) through hCG1 and NUP155 in mammalian cells and in conjunction with inositol hexakisphosphate (IP6), stimulates Dbp5, a member of the DEAD-box helicase family, triggering mRNP remodeling and facilitating RNA export from the nucleus. Recent evidence suggests that mutations in Gle1 causing defects in mRNA export can result in human disease. At least three isoforms of Gle1 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GLE1 RNA export mediator; GLE1 RNA export mediator homolog; GLE1 RNA export mediator-like (yeast; GLE1-like protein; GLE1-like, RNA export mediator; hGLE1; mRNA export factor GLE1; Nucleoporin GLE1
Gene Aliases: 4933405K21Rik; AA553313; GLE1; GLE1L; hGLE1; LCCS; LCCS1
UniProt ID: (Human) Q53GS7, (Mouse) Q8R322, (Rat) Q4KLN4
Entrez Gene ID: (Human) 2733, (Mouse) 74412, (Rat) 362098
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support