Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Expected to cross react with chicken (100% identity), human (100% identity), mouse, (94% identity), Xenopus laevis (100% identity) due to sequence homology.
This gene encodes a protein which belongs to the C2H2-type zinc finger proteins subclass of the Gli family. They are characterized as DNA-binding transcription factors and are mediators of Sonic hedgehog (Shh) signaling. The protein encoded by this gene localizes in the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. Mutations in this gene have been associated with several diseases, including Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, and postaxial polydactyly types A1 and B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); GLI3 form of 190 kDa; GLI3 full length protein; GLI3 full-length protein; GLI3-190; GLI3FL; glioma-associated oncogene family zinc finger 3; neural specific DNA binding protein; Neural-specific DNA-binding protein xGLI3; oncogene GLI3; Transcriptional activator GLI3; xGLI-3; Zinc finger protein GLI3; zinc finger transcription factor Gli3
Gene Aliases: ACLS; GCPS; GLI3; GLI3-190; GLI3FL; im:7142603; PAP-A; PAPA; PAPA1; PAPB; PHS; PPDIV; XELAEV_18031191mg; xgli3
UniProt ID: (Human) P10071, (Chicken) Q9IA31, (Chimpanzee) Q5IS56, (Xenopus) Q91660
Entrez Gene ID: (Dog) 483244, (Human) 2737, (Chicken) 420769, (Chimpanzee) 463369, (Zebrafish) 403042, (Xenopus) 397837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support