Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Positive test controls include: HeLa.
Immunogen sequence: PNESAGSMVN GMQNQDPVGQ GYLAHQLLGD SMQHPGAGRP GQQMLGQISA TSHINIYQGP ESCLPGAHGM GSQPSSLAVV RGYQPCASFG GSRRQAMPRD SLALQSGQLS DTSQTCRVNG IKMEMKGQPH PLCSNLQNYS GQFYDQTVGF SQQDTKAGSF SISDASCLLQ GTSAKNSELL SPGANQVTST VDSLDSHDLE GVQIDFDAII DDGDHSSLMS GALSPSIIQN LSHSSSRLTT PRASLPFPAL SMSTTNMAIG DMSSLLTSLA EESKFLAVMQ
This gene encodes a protein which belongs to the C2H2-type zinc finger proteins subclass of the Gli family. They are characterized as DNA-binding transcription factors and are mediators of Sonic hedgehog (Shh) signaling. The protein encoded by this gene localizes in the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. Mutations in this gene have been associated with several diseases, including Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, and postaxial polydactyly types A1 and B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anterior digit pattern deformity; brachyphalangy; extra toes; GLI-Kruppel family member GLI3; GLI3 form of 190 kDa; GLI3 full length protein; GLI3 full-length protein; GLI3-190; GLI3FL; glioma-associated oncogene family zinc finger 3; oncogene GLI3; polydactyly Nagoya; Transcriptional activator GLI3; zinc finger protein GLI3
Gene Aliases: ACLS; add; AI854843; AU023367; Bph; GCPS; GLI3; GLI3-190; GLI3FL; PAP-A; PAPA; PAPA1; PAPB; Pdn; PHS; PPDIV; Xt
UniProt ID: (Human) O75219, (Mouse) Q61602
Entrez Gene ID: (Human) 2737, (Rat) 140588, (Mouse) 14634
Molecular Function: DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support