Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the glycosyltransferase family. The specific function of this protein has not been determined. Alternative splicing results in multiple transcript variants of this gene [provided by RefSeq, May 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017
Gene Aliases: 2410004H05Rik; 5430414N14Rik; AD-017; AI450005; GALA4A; GLT8D1; MSTP137; MSTP139; UNQ572/PRO1134
UniProt ID: (Human) Q68CQ7, (Mouse) Q6NSU3
Entrez Gene ID: (Human) 55830, (Mouse) 76485
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support