Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MTPGFRDKTF VVQGFGNVGL HSMRYLHRFG AKCIAVGESD GSIWNPDGID PKELEDFKLQ HGSILGFPKA KPYEGSILEV DCDILIPAAT EKQLTKSNAP RVKAKIIAEG ANGPTTPEAD KIFLERNILV IPDLYLNAGG VTVSYFEWLK NLNHVSYGRL TFKYERDSNY HLLLSVQESL ERKFGKHGGT IPIVPTAEFQ DSISGASEKD IVHSALAYTM ERSARQIMHT AMKYNLGLDL RTAAYVNAIE KVFKVYSEAG VTFT
Glutamate dehydrogenase 2, Glud2, is localized to the mitochondrion and acts as a homohexamer to recycle glutamate during neurotransmission. The enzyme catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate. Diseases associated with GLUD2 include Parkinson Disease, Late-Onset and Hyperinsulinemic Hypoglycemia, Familial, 6. Among its related pathways are GABA shunt and Carbon metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDH 2; GluR delta2; Glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; testicular secretory protein Li 14
Gene Aliases: GDH2; GLUD2; GLUDP1
UniProt ID: (Human) P49448
Entrez Gene ID: (Human) 2747
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support