Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Glia maturation factor gamma (GMFG) is a cytokine-responsive protein in erythropoietin-induced and granulocyte-colony stimulating factor-induced hematopoietic lineage development. It may be involved in glial differentiation, neural regeneration, and inhibition of tumor cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glia maturation factor gamma; glia maturation factor, gamma; GMF-gamma
Gene Aliases: 0610039G16Rik; 2310057N07Rik; AI324845; GMF-GAMMA; GMFG
UniProt ID: (Human) O60234, (Rat) Q80T18, (Mouse) Q9ERL7
Entrez Gene ID: (Human) 9535, (Rat) 113940, (Mouse) 63986
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support