Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MLKAVILIGG PQKGTRFRPL SFEVPKPLFP VAGVPMIQHH IEACAQVPGM QEILLIGFYQ PDEPLTQFLE AAQQEFNLPV RYLQEFAPLG TGGGLYHFRD QILAGSPEAF FVLNADVCSD FPLSAMLEAH RRQRHPFLLL GTTANRTQSL NYGCIVENPQ THEVLHYVEK PSTFISDIIN CGIYLFSPEA LKPLRDVFQR NQQDGQLEDS PGLWPGAGTI RLEQDVFSAL AGQGQIYVHL TDGIWSQIKS AGSALYASRL YLSRYQDTHP ERLAKHTPGG PWIRGNVYIH PTAKVAPSAV LGPNVSIGKG VTVGEGVRLR ESIVLHGATL QEHTCVLHSI VGWGSTVGRW ARVEGTPSDP NPNDPRARMD SESLFKDGKL LPAITILGCR VRIPAEVLIL NSIVLPHKEL SRSFTNQIIL
GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDP-mannose pyrophosphorylase A; GMPP-alpha; GTP-mannose-1-phosphate guanylyltransferase alpha; mannose-1-phosphate guanyltransferase alpha; mannose-1-phosphate guanylyltransferase (GDP); Mannose-1-phosphate guanylyltransferase regulatory subunit alpha
Gene Aliases: 1810012N01Rik; AAMR; GMPPA
UniProt ID: (Human) Q96IJ6, (Mouse) Q922H4
Entrez Gene ID: (Human) 29926, (Mouse) 69080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support