Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody can recognize both GMPR1 and GMPR2 due to the high homology.
Immunogen sequence: MPRIDADLK LDFKDVLLRP KRSSLKSRAE VDLERTFTFR NSKQTYSGIP IIVANMDTVG TFEMAAVMSQ HSMFTAIHKH YSLDDWKLFA TNHPECLQNV AVSSGSGQND LEKMTSILEA VPQVKFICLD VANGYSEHFV EFVKLVRAKF PEHTIMAGNV VTGEMVEELI LSGADIIKVG VGPGSVCTTR TKTGVGYPQL SAVIECADSA HGLKGHIISD GGCTCPGDVA KAFGAGADFV MLGGMFSGHT ECAGEVIERN GRKLKLFYGM SSDTAMNKHA GGVAEYRASE GKTVEVPYKG DVENTILDIL GGLRSTCTYV GAAKLKELSR RATFIRVTQQ HNTVFS (1-345 aa encoded by B C008281)
This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GMP reductase 1; GMP reductase 2; GMPR 1; GMPR 2; guanine monophosphate reductase; Guanosine 5'-monophosphate oxidoreductase 1; Guanosine 5'-monophosphate oxidoreductase 2; Guanosine monophosphate reductase 1; Guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog
Gene Aliases: 1810008P16Rik; 2310004P21Rik; 5730544D12Rik; AA959850; AV028449; GMPR; GMPR 1; GMPR 2; GMPR1; GMPR2
UniProt ID: (Human) Q96HQ6, (Human) Q9P2T1, (Mouse) Q9DCZ1, (Rat) Q9Z244, (Mouse) Q99L27
Entrez Gene ID: (Human) 2766, (Human) 51292, (Mouse) 66355, (Rat) 117533, (Mouse) 105446, (Rat) 192357
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support