Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GMP reductase 1; GMPR 1; guanine monophosphate reductase; Guanosine 5'-monophosphate oxidoreductase 1; Guanosine monophosphate reductase 1
Gene Aliases: 2310004P21Rik; AV028449; GMPR; GMPR 1; GMPR1
UniProt ID: (Human) Q96HQ6, (Mouse) Q9DCZ1, (Rat) Q9Z244
Entrez Gene ID: (Human) 2766, (Mouse) 66355, (Rat) 117533
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support