Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPHIDNDVKL DFKDVLLRPK RSTLKSRSEV DLTRSFSFRN SKQTYSGVPI IAANMDTVGT FEMAKVLCKF SLFTAVHKHY SLVQWQEFAG QNPDCLEHLA ASSGTGSSDF EQLEQILEAI PQVKYICLDV ANGYSEHFVE FVKDVRKRFP QHTIMAGNVV TGEMVEELIL SGADIIKVGI GPGSVCTTRK KTGVGYPQLS AVMECADAAH GLKGHIISDG GCSCPGDVAK AFGAGADFVM LGGMLAGHSE SGGELIERDG KKYKLFYGMS SEMAMKKYAG GVAEYRASEG KTVEVPFKGD VEHTIRDILG GIRSTCTYVG AAKLKELSRR TTFIRVTQQV NPIFSEAC
Catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides (PubMed:12009299, PubMed:12669231, PubMed:16359702, PubMed:22037469). Plays a role in modulating cellular differentiation (PubMed:12669231).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GMP reductase 2; GMPR 2; Guanosine 5'-monophosphate oxidoreductase 2; Guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog
Gene Aliases: GMPR 2; GMPR2
UniProt ID: (Human) Q9P2T1
Entrez Gene ID: (Human) 51292
Molecular Function: reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support