Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MADFLPSRSV LSVCFPGCLL TSGEAEQQRK SKEIDKCLSR EKTYVKRLVK ILLLGAGESG KSTFLKQMRI IHGQDFDQRA REEFRPTIYS NVIKGMRVLV DAREKLHIPW GDNSNQQHGD KMMSFDTRAP MAAQGMVETR VFLQYLPAIR ALWADSGIQN AYDRRREFQL GESVKYFLDN LDKLGEPDYI PSQQDILLAR RPTKGIHEYD FEIKNVPFKM VDVGGQRSER KRWFECFDSV TSILFLVSSS EFDQVLMEDR LTNRLTESLN IFETIVNNRV FSNVSIILFL NKTDLLEEKV QIVSIKDYFL EFEGDPHCLR DVQKFLVECF RNKRRDQQQK PLYHHFTTAI NTENIRLVFR DVKDTILHDN LKQLMLQ
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems (PubMed:15240885, PubMed:16787920, PubMed:16705036, PubMed:27084452). Activates effector molecule RhoA by binding and activating RhoGEFs (ARHGEF1/p115RhoGEF, ARHGEF11/PDZ-RhoGEF and ARHGEF12/LARG) (PubMed:15240885, PubMed:12515866). GNA13-dependent Rho signaling subsequently regulates transcription factor AP-1 (activating protein-1). Promotes tumor cell invasion and metastasis by activating RhoA/ROCK signaling pathway (PubMed:16787920, PubMed:16705036, PubMed:27084452). Inhibits CDH1-mediated cell adhesion in process independent from Rho activation (PubMed:11976333). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G alpha 13; G alpha-13; G-protein subunit alpha-13; Galpha 13; guanine nucleotide binding protein (G protein), alpha 13; Guanine nucleotide-binding protein subunit alpha-13
Gene Aliases: G13; GNA13
UniProt ID: (Human) Q14344
Entrez Gene ID: (Human) 10672
Molecular Function:
heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support