Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
GNAI1/GNAI2/GNAI3 are genes that encode for the G protein alpha inhibiting activity subunit. These proteins play a crucial role in signal transduction pathways, particularly in the inhibition of adenylyl cyclase. They are involved in various physiological processes, including neurotransmitter release, hormone secretion, and regulation of smooth muscle contraction. Mutations in these genes have been associated with various diseases, including hypoparathyroidism, pituitary adenoma, and cardiovascular disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G(i) alpha-3; Galpha i1a; Galpha i3; Gi1 protein alpha subunit; GTP-binding protein (G-alpha-i2); GTP-binding regulatory protein Gi alpha-2 chain; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting 3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; guanine nucleotide binding protein, alpha inhibiting 1; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting 3; Guanine nucleotide binding, protein, alpha inhibiting polypeptide 3; Guanine nucleotide-binding protein G(i) subunit alpha-1; Guanine nucleotide-binding protein G(i) subunit alpha-2; Guanine nucleotide-binding protein G(i) subunit alpha-3; guanine nucleotide-binding protein G(i), alpha-1 subunit; guanine nucleotide-binding protein G(i), alpha-2 subunit; guanine nucleotide-binding protein G(k) subunit alpha; HG1B/HG1C/HG1A
Gene Aliases: 87U6; AI158965; ARCND1; AU046200; AW537698; BPGTPB; C76432; Galphai2; Galphai3; Gi; Gia; Gialpha1; GIP; GIP3A; Gnai-1; Gnai-2; Gnai-3; GNAI1; GNAI2; GNAI2B; GNAI3; H_LUCA15.1; H_LUCA16.1; zgc:63957
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support