Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cone-type transducin alpha subunit; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; Guanine nucleotide-binding protein G(t) subunit alpha-2; RP5-1160K1.4; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide
Gene Aliases: ACHM4; AW490837; Gnat-2; GNAT2; GNATC; Gt-2; Tcalpha
UniProt ID: (Human) P19087, (Mouse) P50149
Entrez Gene ID: (Human) 2780, (Mouse) 14686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support