Search Thermo Fisher Scientific
PA1-27694 detects GNB4 from human and mouse samples.
PA1-27694 may be successfully used in Western blot applications.
The PA1-27719 immunogen is a synthetic peptide corresponding to human Guanine nucleotide-binding protein subunit beta 4.
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors.