Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific for a peptide sequence within the D1 blade which is not found in the Gb's 1-4.
Immunogen sequence: MATEGLHEN ETLASLKSEA ESLKGKLEEE RAKLHDVELH QVAERVEALG QFVMKTRRTL KGHGNKVLCM DWCKDKRRIV SSSQDGKVIV WDSFTTNKVR HCSQPRYRGF RIPAYLKVLW SSCPALS (42-140 aa encoded by BC011671)
The Gb5 protein is a G protein subunit present predominantly in the central nervous system. This protein appears to be expressed in two forms, one found in the brain of 39 kDa and one found in the retina of 44 kDa. It has also been found that Gb5 associates tightly with RGS7, a member of the superfamily of negative regulators of G protein signaling. These complexes have been localized to most regions of the brain, whereas Gb1-4 exhibit no observable colocalization with RGS7, despite their widespread codistribution throughout similar neuronal networks. This antibody is specific for a peptide sequence within the D1 blade which is not found in the Gb's 1-4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: flail; flailer; G protein, beta subunit 5L; G protein, beta-5 subunit; GBB5; Gbeta5; guanine nucleotide binding protein (G protein), beta 5; guanine nucleotide binding protein (G protein), beta polypeptide 5; guanine nucleotide binding protein, beta 5; Guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; guanine nucleotide-binding protein, beta-5 subunit; hug; Transducin beta chain 5
Gene Aliases: flr; GB5; Gbeta5; GBS; GNB5
UniProt ID: (Human) Q9HAU9, (Mouse) P62881, (Rat) P62882
Entrez Gene ID: (Human) 10681, (Mouse) 14697, (Rat) 83579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support