Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Antibody detects endogenous levels of total GNG3.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: guanine nucleotide binding protein (G protein), gamma 3; guanine nucleotide binding protein (G protein), gamma 3 subunit; guanine nucleotide binding protein, gamma 3; Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-3; guanine nucleotide-binding protein gamma-3 subunit; NBP gamma-3
Gene Aliases: GNG3; GNGT3
UniProt ID: (Human) P63215, (Mouse) P63216
Entrez Gene ID: (Human) 2785, (Mouse) 14704, (Rat) 114117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support