Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKEGMSNNS TTSISQARKA VEQLKMEACM DRVKVSQAAA DLLAYCEAHV REDPLIIPVP ASENPFREKK FFCTIL (1-75 aa encoded by BC022485)
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: guanine nucleotide binding protein (G protein), gamma 4; guanine nucleotide binding protein (G protein), gamma 4 subunit; guanine nucleotide binding protein 4; Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-4
Gene Aliases: GNG4; GNGT4
UniProt ID: (Human) P50150, (Mouse) P50153
Entrez Gene ID: (Human) 2786, (Mouse) 14706, (Rat) 114118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support