Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA:dihydroxyacetonephosphateacyltransferase; DAP-AT; DHAP-AT; Dihydroxyacetone phosphate acyltransferase; Glycerone-phosphate O-acyltransferase
Gene Aliases: DAP-AT; DAPAT; DHAPAT; GNPAT; RCDP2
UniProt ID: (Human) O15228
Entrez Gene ID: (Human) 8443
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support