Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKLIILEHY SQASEWAAKY IRNRIIQFNP GPEKYFTLGL PTGSTPLGCY KKLIEYYKNG DLSFKYVKTF NMDEYVGLPR DHPESYHSFM WNNFFKHIDI HPENTHILDG NAVDLQAECD AFEEKIKAAG GIELFVGGIG PDGHIAFNEP GSSLVSRTRV KTLAMDTILA NARFFDGELT KVPTMALTVG VGTVMDAREV MILITGAHKA FALYKAIEEG VNHMWTVSAF QQHPRTVFVC DEDATLELKV KTVKYFKGLM LVHNKLVDPL YSIKEKETEK SQSSKKPYSD (1-289 aa encoded by BC012853)
GNPDA1 is a 289 amino acid cytoplasmic protein belonging to the glucosamine/ galactosamine-6-phosphate isomerase family. It is one of the two mammalian glucosamine-6-phosphate deaminase enzymes. GNPDA1 is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate (GlcN6P) into D-fructose-6-phosphate (Fru6P) and ammonium with an aldo/keto isomerization and an amination/deamination. GNPDA1 may trigger calcium oscillations in mammalian eggs at fertilization which in turn serve as an important factor for egg development and early embryo development. It is a homohexamer, ubiquitously expressed in most of human tissues with high expression in testis, ovary, placenta, and heart and also in various cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glcN6P deaminase 1; Glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; GNPDA 1; Oscillin
Gene Aliases: GNP1; GNPDA; GNPDA1; GNPI; GPI; HLN; KIAA0060; oscillin
UniProt ID: (Human) P46926, (Mouse) O88958
Entrez Gene ID: (Human) 10007, (Mouse) 26384, (Rat) 683570
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support