Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKPDETPMF DPSLLKEVDW SQNTATFSPA ISPTHPGEGL VLRPLCTADL NRGFFKVLGQ LTETGVVSPE QFMKSFEHMK KSGDYYVTVV EDVTLGQIVA TATLIIEHKF IHSCAKRGRV EDVVVSDECR GKQLGKLLLS TLTLLSKKLN CYKITLECLP QNVGFYKKFG YTVSEENYMC RRFLK (1-184 aa encoded by BC012179)
Glucosamine-6 phosphate-N-acetyl Transferase (GNPNAT1) is a key enzyme in the hexosamine biosynthetic pathway. It converts D-glucosamine 6-phosphate to N-acetyl-D-glucosamine 6-phosphate, the acetylation step allowing carbon and nitrogen enter into the hexosamine biosynthetic pathway. Recently inhibition of GNPNAT1 has been reported to promote proliferation and aggressiveness of castration-resistant prostate cancer. (27194471).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glucosamine 6-phosphate N-acetyltransferase; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase 1; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase; Phosphoglucosamine acetylase; Phosphoglucosamine transacetylase; Protein EMeg32
Gene Aliases: AU017428; AU040593; EMeg32; GNA1; GNPNAT; GNPNAT1; Gpnat1; Gsnpat; RGD1563144
UniProt ID: (Human) Q96EK6, (Mouse) Q9JK38
Entrez Gene ID: (Human) 64841, (Mouse) 54342, (Rat) 498486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support