Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRLLPLAPGR LRRGSPRHLP SCSPALLLLV LGGCLGVFGV AAGTRRPNVV LLLTDDQDEV LGGMTPLKKT KALIGEMGMT FSSAYVPSAL CCPSRASILT GKYPHNHHVV NNTLEGNCSS KSWQKIQEPN TFPAILRSMC GYQTFFAGKY LNEYGAPDAG GLEHVPLGWS YWYALEKNSK YYNYTLSING KARKHGENYS VDYLTDVLAN VSLDFLDYKS NFEPFFMMIA TPAPHSPWTA APQYQKAFQN VFAPRNKNFN IHGTNKHWLI RQAKTPMTNS SIQFLDNAFR KRWQTLLSVD DLVEKLVKRL EFTGELNNTY IFYTSDNGYH TGQFSLPIDK RQLYEFDIKV PLLVRGPGIK PNQTSKMLVA NIDLGPTILD IAGYDLNKTQ MDGMSLLPIL RGASNLTWRS DVLVEYQGEG RNVTDPTCPS LSPGVSQCFP DCVCEDAYNN TYACVRTMSA LWNLQYCEFD DQEVFVEVYN LTADPDQITN IAKTIDPELL GKMNYRLMML QSCSGPTCRT PGVFDPGYRF DPRLMFSNRG SVRTRRFSKH LL
GNS is a lysosomal enzyme found in all cells. It is involved in the catabolism of heparin, heparan sulphate, and keratan sulphate. Deficiency of this enzyme results in the accumulation of undegraded substrate and the lysosomal storage disorder mucopolysaccharidosis type IIID (Sanfilippo D syndrome). Mucopolysaccharidosis type IIID is the least common of the four subtypes of Sanfilippo syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G6S; glucosamine -6-sulfatase; Glucosamine-6-sulfatase; N-acetylglucosamine-6-sulfatase
Gene Aliases: G6S; GNS
UniProt ID: (Human) P15586
Entrez Gene ID: (Human) 2799
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support