Product References
SARS-CoV-2 accessory proteins ORF7a and ORF3a use distinct mechanisms to down-regulate MHC-I surface expression.
Proceedings of the National Academy of Sciences of the United States of America
Arshad N,Laurent-Rolle M,Ahmed WS,Hsu JC,Mitchell SM,Pawlak J,Sengupta D,Biswas KH,Cresswell P
Tue Jan 03 00:00:00 UTC 2023
Lanthanum Chloride Induces Axon Abnormality Through LKB1-MARK2 and LKB1-STK25-GM130 Signaling Pathways.
Cellular and molecular neurobiology
Song Z,Mao H,Liu J,Sun W,Wu S,Lu X,Jin C,Yang J
Sat Apr 01 00:00:00 UTC 2023
Intracellular Vimentin Regulates the Formation of Classical Swine Fever Virus Replication Complex through Interaction with NS5A Protein.
Journal of virology
Cheng Y,Lou JX,Liu YY,Liu CC,Chen J,Yang MC,Ye YB,Go YY,Zhou B
Wed May 31 00:00:00 UTC 2023
Exosomes derived from oral squamous cell carcinoma tissue accelerate diabetic wound healing.
American journal of physiology. Cell physiology
Zhang M,Guo J,Xiang K,Chen J,Wang C,Jiang T,Kang Y,Xu X,Li J,Yang X,Chen Z
Thu Jun 01 00:00:00 UTC 2023
SLC35A2 deficiency reduces protein levels of core 1 β-1,3-galactosyltransferase 1 (C1GalT1) and its chaperone Cosmc and affects their subcellular localization.
Biochimica et biophysica acta. Molecular cell research
Wiertelak W,Chabowska K,Szulc B,Zadorozhna Y,Olczak M,Maszczak-Seneczko D
Thu Jun 01 00:00:00 UTC 2023
Chemokine receptor CXCR7 activates Aurora Kinase A and promotes neuroendocrine prostate cancer growth.
The Journal of clinical investigation
Gritsina G,Fong KW,Lu X,Lin Z,Xie W,Agarwal S,Lin D,Schiltz GE,Beltran H,Corey E,Morrissey C,Wang Y,Zhao JC,Hussain M,Yu J
Tue Aug 01 00:00:00 UTC 2023
Golgi damage caused by dysfunction of PiT-2 in primary familial brain calcification.
Biochemical and biophysical research communications
Sun H,Wang Z,Zhang Q,Chen N,Tang M,Yang Z,Xu Y,Kang J,Wang Y
Sun Jan 29 00:00:00 UTC 2023
Golgi damage caused by dysfunction of PiT-2 in primary familial brain calcification.
Biochemical and biophysical research communications
Sun H,Wang Z,Zhang Q,Chen N,Tang M,Yang Z,Xu Y,Kang J,Wang Y
Sun Jan 29 00:00:00 UTC 2023
New Loss-of-Function Mutations in PCSK9 Reduce Plasma LDL Cholesterol.
Arteriosclerosis, thrombosis, and vascular biology
Meng FH,Liu S,Xiao J,Zhou YX,Dong LW,Li YF,Zhang YQ,Li WH,Wang JQ,Wang Y,Song BL,Ma YT,Fu ZY,Luo J
Sat Jul 01 00:00:00 UTC 2023
BMSC-derived extracellular vesicles promoted osteogenesis via Axin2 inhibition by delivering MiR-16-5p.
International immunopharmacology
Duan J,Li H,Wang C,Yao J,Jin Y,Zhao J,Zhang Y,Liu M,Sun H
Sat Jul 01 00:00:00 UTC 2023
Hydrogel armed with Bmp2 mRNA-enriched exosomes enhances bone regeneration.
Journal of nanobiotechnology
Yang Z,Li X,Gan X,Wei M,Wang C,Yang G,Zhao Y,Zhu Z,Wang Z
Wed Apr 05 00:00:00 UTC 2023
Gβγ signaling regulates microtubule-dependent control of Golgi integrity.
Cellular signalling
Rajanala K,Wedegaertner PB
Thu Jun 01 00:00:00 UTC 2023
Exosomal RBP4 potentiated hepatic lipid accumulation and inflammation in high-fat-diet-fed mice by promoting M1 polarization of Kupffer cells.
Free radical biology & medicine
Yao JM,Ying HZ,Zhang HH,Qiu FS,Wu JQ,Yu CH
Wed Feb 01 00:00:00 UTC 2023
ERK and USP5 govern PD-1 homeostasis via deubiquitination to modulate tumor immunotherapy.
Nature communications
Xiao X,Shi J,He C,Bu X,Sun Y,Gao M,Xiang B,Xiong W,Dai P,Mao Q,Xing X,Yao Y,Yu H,Xu G,Li S,Ren Y,Chen B,Jiang C,Meng G,Lee YR,Wei W,Freeman GJ,Xie C,Zhang J
Fri May 19 00:00:00 UTC 2023
ERK and USP5 govern PD-1 homeostasis via deubiquitination to modulate tumor immunotherapy.
Nature communications
Xiao X,Shi J,He C,Bu X,Sun Y,Gao M,Xiang B,Xiong W,Dai P,Mao Q,Xing X,Yao Y,Yu H,Xu G,Li S,Ren Y,Chen B,Jiang C,Meng G,Lee YR,Wei W,Freeman GJ,Xie C,Zhang J
Fri May 19 00:00:00 UTC 2023
hASCs-derived exosomal miR-155-5p targeting TGFβR2 promotes autophagy and reduces pyroptosis to alleviate intervertebral disc degeneration.
Journal of orthopaedic translation
Chen D,Jiang X,Zou H
Wed Mar 01 00:00:00 UTC 2023
Lipid phosphatase SAC1 suppresses hepatitis B virus replication through promoting autophagic degradation of virions.
Antiviral research
Zheng J,Deng Y,Wei Z,Zou H,Wen X,Cai J,Zhang S,Jia B,Lu M,Lu K,Lin Y
Mon May 01 00:00:00 UTC 2023
Tumor tissue derived extracellular vesicles promote diabetic wound healing.
Journal of diabetes and its complications
Guo J,Jiang G,Chen J,Zhang M,Xiang K,Wang C,Jiang T,Kang Y,Sun Y,Xu X,Yang X,Chen Z
Mon May 01 00:00:00 UTC 2023
FOXP3+ macrophage represses acute ischemic stroke-induced neural inflammation.
Autophagy
Cai W,Hu M,Li C,Wu R,Lu D,Xie C,Zhang W,Li T,Shen S,Huang H,Qiu W,Liu Q,Lu Y,Lu Z
Sat Apr 01 00:00:00 UTC 2023
Exercise promotes angiogenesis by enhancing endothelial cell fatty acid utilization via liver-derived extracellular vesicle miR-122-5p.
Journal of sport and health science
Lou J,Wu J,Feng M,Dang X,Wu G,Yang H,Wang Y,Li J,Zhao Y,Shi C,Liu J,Zhao L,Zhang X,Gao F
Fri Jul 01 00:00:00 UTC 2022
Exosomes from Adipose Stem Cells Promote Diabetic Wound Healing through the eHSP90/LRP1/AKT Axis.
Cells
Ren S,Chen J,Guo J,Liu Y,Xiong H,Jing B,Yang X,Li G,Kang Y,Wang C,Xu X,Liu Z,Zhang M,Xiang K,Li C,Li Q,Machens HG,Chen Z
Fri Oct 14 00:00:00 UTC 2022
Macrophages with Different Polarization Phenotypes Influence Cementoblast Mineralization through Exosomes.
Stem cells international
Zhao Y,Huang Y,Liu H,Tan K,Wang R,Jia L,Li W
Wed Sep 28 00:00:00 UTC 2022
N-Glycosylation on Asn50 of SND1 Is Required for Glioma U87 Cell Proliferation and Metastasis.
Journal of immunology research
Zhou Y,Li Q,Zheng J,Lin N
Tue Oct 11 00:00:00 UTC 2022
Bunyavirus SFTSV exploits autophagic flux for viral assembly and egress.
Autophagy
Yan JM,Zhang WK,Yan LN,Jiao YJ,Zhou CM,Yu XJ
Fri Jul 01 00:00:00 UTC 2022
Bunyavirus SFTSV exploits autophagic flux for viral assembly and egress.
Autophagy
Yan JM,Zhang WK,Yan LN,Jiao YJ,Zhou CM,Yu XJ
Fri Jul 01 00:00:00 UTC 2022
Bunyavirus SFTSV exploits autophagic flux for viral assembly and egress.
Autophagy
Yan JM,Zhang WK,Yan LN,Jiao YJ,Zhou CM,Yu XJ
Fri Jul 01 00:00:00 UTC 2022
Bunyavirus SFTSV exploits autophagic flux for viral assembly and egress.
Autophagy
Yan JM,Zhang WK,Yan LN,Jiao YJ,Zhou CM,Yu XJ
Fri Jul 01 00:00:00 UTC 2022
ER proteins decipher the tubulin code to regulate organelle distribution.
Nature
Zheng P,Obara CJ,Szczesna E,Nixon-Abell J,Mahalingan KK,Roll-Mecak A,Lippincott-Schwartz J,Blackstone C
Sat Jan 01 00:00:00 UTC 2022
LMAN1-MCFD2 complex is a cargo receptor for the ER-Golgi transport of α1-antitrypsin.
The Biochemical journal
Zhang Y,Zhu M,Zheng C,Wei W,Emmer BT,Zhang B
Thu Apr 14 00:00:00 UTC 2022
SARS-CoV-2 Causes Lung Inflammation through Metabolic Reprogramming and RAGE.
Viruses
Allen CNS,Santerre M,Arjona SP,Ghaleb LJ,Herzi M,Llewellyn MD,Shcherbik N,Sawaya BE
Fri May 06 00:00:00 UTC 2022
"All-in-One" Exosome Engineering Strategy for Effective Therapy of Familial Hypercholesterolemia.
ACS applied materials & interfaces
Mei R,Wan Z,Li Z,Wei M,Qin W,Yuan L,Liu L,Yang G
Wed Nov 16 00:00:00 UTC 2022
Exercise-derived peptide protects against pathological cardiac remodeling.
EBioMedicine
Yin A,Yuan R,Xiao Q,Zhang W,Xu K,Yang X,Yang W,Xu L,Wang X,Zhuang F,Li Y,Cai Z,Sun Z,Zhou B,He B,Shen L
Mon Aug 01 00:00:00 UTC 2022
Sonodynamical reversion of immunosuppressive microenvironment in prostate cancer via engineered exosomes.
Drug delivery
Wang D,Wan Z,Yang Q,Chen J,Liu Y,Lu F,Tang J
Thu Dec 01 00:00:00 UTC 2022
GALNT1 Enhances Malignant Phenotype of Gastric Cancer via Modulating CD44 Glycosylation to Activate the Wnt/β-catenin Signaling Pathway.
International journal of biological sciences
Zhang J,Wang H,Wu J,Yuan C,Chen S,Liu S,Huo M,Zhang C,He Y
Tue Nov 29 00:00:00 UTC 2022
Characterization and subcellular localization of Alongshan virus proteins.
Frontiers in microbiology
Zhao Y,Wu P,Liu L,Ma B,Pan M,Huang Y,Du N,Yu H,Sui L,Wang ZD,Hou Z,Liu Q
Sat Oct 15 00:00:00 UTC 2022
TissUExM enables quantitative ultrastructural analysis in whole vertebrate embryos by expansion microscopy.
Cell reports methods
Steib E,Tetley R,Laine RF,Norris DP,Mao Y,Vermot J
Mon Oct 24 00:00:00 UTC 2022
Selenoprotein K contributes to CD36 subcellular trafficking in hepatocytes by accelerating nascent COPII vesicle formation and aggravates hepatic steatosis.
Redox biology
You M,Wu F,Gao M,Chen M,Zeng S,Zhang Y,Zhao W,Li D,Wei L,Ruan XZ,Chen Y
Tue Nov 01 00:00:00 UTC 2022
Selenoprotein K contributes to CD36 subcellular trafficking in hepatocytes by accelerating nascent COPII vesicle formation and aggravates hepatic steatosis.
Redox biology
You M,Wu F,Gao M,Chen M,Zeng S,Zhang Y,Zhao W,Li D,Wei L,Ruan XZ,Chen Y
Tue Nov 01 00:00:00 UTC 2022
Selenoprotein K contributes to CD36 subcellular trafficking in hepatocytes by accelerating nascent COPII vesicle formation and aggravates hepatic steatosis.
Redox biology
You M,Wu F,Gao M,Chen M,Zeng S,Zhang Y,Zhao W,Li D,Wei L,Ruan XZ,Chen Y
Tue Nov 01 00:00:00 UTC 2022
Selenoprotein K contributes to CD36 subcellular trafficking in hepatocytes by accelerating nascent COPII vesicle formation and aggravates hepatic steatosis.
Redox biology
You M,Wu F,Gao M,Chen M,Zeng S,Zhang Y,Zhao W,Li D,Wei L,Ruan XZ,Chen Y
Tue Nov 01 00:00:00 UTC 2022
Visualizing the native cellular organization by coupling cryofixation with expansion microscopy (Cryo-ExM).
Nature methods
Laporte MH,Klena N,Hamel V,Guichard P
Tue Feb 01 00:00:00 UTC 2022
African Swine Fever Virus K205R Induces ER Stress and Consequently Activates Autophagy and the NF-κB Signaling Pathway.
Viruses
Wang Q,Zhou L,Wang J,Su D,Li D,Du Y,Yang G,Zhang G,Chu B
Tue Feb 15 00:00:00 UTC 2022
SARS-CoV-2 accessory proteins ORF7a and ORF3a use distinct mechanisms to downregulate MHC-I surface expression.
bioRxiv : the preprint server for biology
Arshad N,Laurent-Rolle M,Ahmed WS,Hsu JC,Mitchell SM,Pawlak J,Sengupta D,Biswas KH,Cresswell P
Tue May 17 00:00:00 UTC 2022
Comparison of the Variability of Small Extracellular Vesicles Derived from Human Liver Cancer Tissues and Cultured from the Tissue Explants Based on a Simple Enrichment Method.
Stem cell reviews and reports
Chen J,Jiao Z,Mo J,Huang D,Li Z,Zhang W,Yang T,Zhao M,Xie F,Hu D,Wang X,Yi X,Jiang Y,Zhong T
Tue Mar 01 00:00:00 UTC 2022
A cryostat-based frozen section method to increase the yield of extracellular vesicles extracted from different tissues.
BioTechniques
Yang S,Qin W,Li X,Guo Q,Wang Y,Chen J,Li H,Zhang S,Min L
Mon Aug 01 00:00:00 UTC 2022
Aberrant Cholesterol Metabolism and Wnt/β-Catenin Signaling Coalesce via Frizzled5 in Supporting Cancer Growth.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zheng S,Lin J,Pang Z,Zhang H,Wang Y,Ma L,Zhang H,Zhang X,Chen M,Zhang X,Zhao C,Qi J,Cao L,Wang M,He X,Sheng R
Sat Oct 01 00:00:00 UTC 2022
c-Abl kinase-mediated phosphorylation of γ-tubulin promotes γ-tubulin ring complexes assembly and microtubule nucleation.
The Journal of biological chemistry
Wang GF,Dong Q,Bai Y,Gu J,Tao Q,Yue J,Zhou R,Niu X,Zhu L,Song C,Zheng T,Wang D,Jin Y,Liu H,Cao C,Liu X
Fri Apr 01 00:00:00 UTC 2022
Exosomes from Microvascular Endothelial Cells under Mechanical Unloading Inhibit Osteogenic Differentiation via miR-92b-3p/ELK4 Axis.
Journal of personalized medicine
Zhang X,Zhang L,Xu L,Li G,Wang K,Xue T,Sun Q,Tang H,Cao X,Hu Z,Zhang S,Shi F
Thu Dec 08 00:00:00 UTC 2022
Neural stem cell‑derived exosomes transfer miR‑124‑3p into cells to inhibit glioma growth by targeting FLOT2.
International journal of oncology
Qian C,Wang Y,Ji Y,Chen D,Wang C,Zhang G,Wang Y
Sat Oct 01 00:00:00 UTC 2022
TM9SF4 is an F-actin disassembly factor that promotes tumor progression and metastasis.
Nature communications
Meng Z,Li Z,Xie M,Yu H,Jiang L,Yao X
Thu Sep 29 00:00:00 UTC 2022
An mTORC1-GRASP55 signaling axis controls unconventional secretion to reshape the extracellular proteome upon stress.
Molecular cell
Nüchel J,Tauber M,Nolte JL,Mörgelin M,Türk C,Eckes B,Demetriades C,Plomann M
Thu Aug 19 00:00:00 UTC 2021
Smart exosomes with lymph node homing and immune-amplifying capacities for enhanced immunotherapy of metastatic breast cancer.
Molecular therapy. Nucleic acids
Ji P,Yang Z,Li H,Wei M,Yang G,Xing H,Li Q
Fri Dec 03 00:00:00 UTC 2021
Exosome-mediated delivery of inflammation-responsive Il-10 mRNA for controlled atherosclerosis treatment.
Theranostics
Bu T,Li Z,Hou Y,Sun W,Zhang R,Zhao L,Wei M,Yang G,Yuan L
Mon Feb 07 00:00:00 UTC 2022
Identification of TRAMs as sphingolipid-binding proteins using a photoactivatable and clickable short-chain ceramide analog.
The Journal of biological chemistry
Deng Y,You L,Lu Y,Han S,Wang J,Vicas N,Chen C,Ye J
Wed Dec 01 00:00:00 UTC 2021
Cell-specific expression of functional glucose transporter 8 in mammary gland.
Biochemical and biophysical research communications
Mardones L,Muñoz K,Villagrán M
Fri Aug 27 00:00:00 UTC 2021
Human umbilical vein endothelial cells derived-exosomes promote osteosarcoma cell stemness by activating Notch signaling pathway.
Bioengineered
Yang J,Hu Y,Wang L,Sun X,Yu L,Guo W
Wed Dec 01 00:00:00 UTC 2021
EVs Containing Host Restriction Factor IFITM3 Inhibited ZIKV Infection of Fetuses in Pregnant Mice through Trans-placenta Delivery.
Molecular therapy : the journal of the American Society of Gene Therapy
Zou X,Yuan M,Zhang T,Zheng N,Wu Z
Wed Jan 06 00:00:00 UTC 2021
Gβγ regulates mitotic Golgi fragmentation and G2/M cell cycle progression.
Molecular biology of the cell
Rajanala K,Klayman LM,Wedegaertner PB
Fri Oct 01 00:00:00 UTC 2021
Receptor-Mediated ER Export of Lipoproteins Controls Lipid Homeostasis in Mice and Humans.
Cell metabolism
Wang X,Wang H,Xu B,Huang D,Nie C,Pu L,Zajac GJM,Yan H,Zhao J,Shi F,Emmer BT,Lu J,Wang R,Dong X,Dai J,Zhou W,Wang C,Gao G,Wang Y,Willer C,Lu X,Zhu Y,Chen XW
Tue Feb 02 00:00:00 UTC 2021
Extracellular vesicles-based pre-targeting strategy enables multi-modal imaging of orthotopic colon cancer and image-guided surgery.
Journal of nanobiotechnology
Jing B,Qian R,Jiang D,Gai Y,Liu Z,Guo F,Ren S,Gao Y,Lan X,An R
Sat May 22 00:00:00 UTC 2021
TMEM41B acts as an ER scramblase required for lipoprotein biogenesis and lipid homeostasis.
Cell metabolism
Huang D,Xu B,Liu L,Wu L,Zhu Y,Ghanbarpour A,Wang Y,Chen FJ,Lyu J,Hu Y,Kang Y,Zhou W,Wang X,Ding W,Li X,Jiang Z,Chen J,Zhang X,Zhou H,Li JZ,Guo C,Zheng W,Zhang X,Li P,Melia T,Reinisch K,Chen XW
Tue Aug 03 00:00:00 UTC 2021
TMEM41B acts as an ER scramblase required for lipoprotein biogenesis and lipid homeostasis.
Cell metabolism
Huang D,Xu B,Liu L,Wu L,Zhu Y,Ghanbarpour A,Wang Y,Chen FJ,Lyu J,Hu Y,Kang Y,Zhou W,Wang X,Ding W,Li X,Jiang Z,Chen J,Zhang X,Zhou H,Li JZ,Guo C,Zheng W,Zhang X,Li P,Melia T,Reinisch K,Chen XW
Tue Aug 03 00:00:00 UTC 2021
Angiogenesis is promoted by exosomal DPP4 derived from 5-fluorouracil-resistant colon cancer cells.
Cancer letters
Zheng X,Liu J,Li X,Tian R,Shang K,Dong X,Cao B
Thu Jan 28 00:00:00 UTC 2021
Identification of interaction domains in the pseudorabies virus ribonucleotide reductase large and small subunits.
Veterinary microbiology
Lyu C,Li WD,Peng JM,Cai XH
Wed Jul 01 00:00:00 UTC 2020
CBS homogenization mutation strategy narrows the glycan binding profile of a GlcNAc-specific lectin AANL.
Glycobiology
Su Y,Ye X,Xu B,Li Y,Yang Q,Yu W,Song J,Guo C,Wang X,Hart GW,Sun H
Wed Feb 19 00:00:00 UTC 2020
Fusion protein engineered exosomes for targeted degradation of specific RNAs in lysosomes: a proof-of-concept study.
Journal of extracellular vesicles
Li Z,Zhou X,Gao X,Bai D,Dong Y,Sun W,Zhao L,Wei M,Yang X,Yang G,Yuan L
Sun Sep 06 00:00:00 UTC 2020
Exosome-mediated Hic-5 regulates proliferation and apoptosis of osteosarcoma via Wnt/β-catenin signal pathway.
Aging
Sha L,Ma D,Chen C
Sun Dec 13 00:00:00 UTC 2020
Loss of GM130 does not impair oocyte meiosis and embryo development in mice.
Biochemical and biophysical research communications
Jiang Y,Liu Y,Han F,Zhou J,Zhang X,Xu J,Yu Z,Zhao S,Gao F,Zhao H
Thu Nov 12 00:00:00 UTC 2020
Loss of GM130 does not impair oocyte meiosis and embryo development in mice.
Biochemical and biophysical research communications
Jiang Y,Liu Y,Han F,Zhou J,Zhang X,Xu J,Yu Z,Zhao S,Gao F,Zhao H
Thu Nov 12 00:00:00 UTC 2020
Mast cells-derived MiR-223 destroys intestinal barrier function by inhibition of CLDN8 expression in intestinal epithelial cells.
Biological research
Li M,Zhao J,Cao M,Liu R,Chen G,Li S,Xie Y,Xie J,Cheng Y,Huang L,Su M,Xu Y,Zheng M,Zou K,Geng L,Xu W,Gong S
Tue Mar 24 00:00:00 UTC 2020
Liquid Biopsy of Extracellular Vesicle-Derived miR-193a-5p in Colorectal Cancer and Discovery of Its Tumor-Suppressor Functions.
Frontiers in oncology
Wei R,Chen L,Qin D,Guo Q,Zhu S,Li P,Min L,Zhang S
Tue Oct 06 00:00:00 UTC 2020
Improved secretion of glycoproteins using an N-glycan-restricted passport sequence tag recognized by cargo receptor.
Nature communications
Yagi H,Yagi-Utsumi M,Honda R,Ohta Y,Saito T,Nishio M,Ninagawa S,Suzuki K,Anzai T,Kamiya Y,Aoki K,Nakanishi M,Satoh T,Kato K
Fri Mar 13 00:00:00 UTC 2020
BMSCs-Derived Exosomes Ameliorate Pain Via Abrogation of Aberrant Nerve Invasion in Subchondral Bone in Lumbar Facet Joint Osteoarthritis.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Li J,Ding Z,Li Y,Wang W,Wang J,Yu H,Liu A,Miao J,Chen S,Wu T,Cao Y
Sun Mar 01 00:00:00 UTC 2020
MACF1 promotes preosteoblast migration by mediating focal adhesion turnover through EB1.
Biology open
Su P,Yin C,Li D,Yang C,Wang X,Pei J,Tian Y,Qian A
Tue Mar 24 00:00:00 UTC 2020
Exosomes derived from 5-fluorouracil-resistant colon cancer cells are enriched in GDF15 and can promote angiogenesis.
Journal of Cancer
Zheng X,Ma N,Wang X,Hu J,Ma X,Wang J,Cao B
Tue Nov 17 00:00:00 UTC 2020
Host BAG3 Is Degraded by Pseudorabies Virus pUL56 C-Terminal 181L-185L and Plays a Negative Regulation Role during Viral Lytic Infection.
International journal of molecular sciences
Lyu C,Li WD,Wang SW,Peng JM,Yang YB,Tian ZJ,Cai XH
Wed Apr 29 00:00:00 UTC 2020
Host BAG3 Is Degraded by Pseudorabies Virus pUL56 C-Terminal 181L-185L and Plays a Negative Regulation Role during Viral Lytic Infection.
International journal of molecular sciences
Lyu C,Li WD,Wang SW,Peng JM,Yang YB,Tian ZJ,Cai XH
Wed Apr 29 00:00:00 UTC 2020
Identification and Verification of Two Novel Differentially Expressed Proteins from Non-neoplastic Mucosa and Colorectal Carcinoma Via iTRAQ Combined with Liquid Chromatography-Mass Spectrometry.
Pathology oncology research : POR
Bai Y,Wang J,Gao Z,Dai E
Wed Apr 01 00:00:00 UTC 2020
Genetically Encoded FapR-NLuc as a Biosensor to Determine Malonyl-CoA in Situ at Subcellular Scales.
Bioconjugate chemistry
Du Y,Hu H,Pei X,Du K,Wei T
Wed Mar 20 00:00:00 UTC 2019
Genetically Encoded FapR-NLuc as a Biosensor to Determine Malonyl-CoA in Situ at Subcellular Scales.
Bioconjugate chemistry
Du Y,Hu H,Pei X,Du K,Wei T
Wed Mar 20 00:00:00 UTC 2019
A GFP-tagged version of the pseudorabies virus protein UL56 localizes to the Golgi and trans-Golgi network through a predicted C-terminal leucine-rich helix in transfected cells.
Virology journal
Lyu C,Cai X
Thu Jun 20 00:00:00 UTC 2019
Golgi apparatus fragmentation participates in oxidized low-density lipoprotein-induced endothelial cell injury.
Journal of cellular biochemistry
Ji G,Song X,Wang L,Li Z,Wu H,Dong H
Fri Nov 01 00:00:00 UTC 2019
Exosomal miR-1229 derived from colorectal cancer cells promotes angiogenesis by targeting HIPK2.
International journal of biological macromolecules
Hu HY,Yu CH,Zhang HH,Zhang SZ,Yu WY,Yang Y,Chen Q
Mon Jul 01 00:00:00 UTC 2019
Exosomes from human adipose-derived stem cells promote sciatic nerve regeneration via optimizing Schwann cell function.
Journal of cellular physiology
Chen J,Ren S,Duscher D,Kang Y,Liu Y,Wang C,Yuan M,Guo G,Xiong H,Zhan P,Wang Y,Machens HG,Chen Z
Sun Dec 01 00:00:00 UTC 2019
Antitumor Potential of Extracellular Vesicles Released by Genetically Modified Murine Colon Carcinoma Cells With Overexpression of Interleukin-12 and shRNA for TGF-β1.
Frontiers in immunology
Rossowska J,Anger N,Wegierek K,Szczygieł A,Mierzejewska J,Milczarek M,Szermer-Olearnik B,Pajtasz-Piasecka E
Thu Jan 16 00:00:00 UTC 2020
Tumor-derived exosomal miRNA-320d as a biomarker for metastatic colorectal cancer.
Journal of clinical laboratory analysis
Tang Y,Zhao Y,Song X,Song X,Niu L,Xie L
Fri Nov 01 00:00:00 UTC 2019
Extracellular vesicles expressing a single-chain variable fragment of an HIV-1 specific antibody selectively target Env+ tissues.
Theranostics
Zou X,Yuan M,Zhang T,Wei H,Xu S,Jiang N,Zheng N,Wu Z
Tue Aug 18 00:00:00 UTC 2020
Impaired Bone Regenerative Effect of Exosomes Derived from Bone Marrow Mesenchymal Stem Cells in Type 1 Diabetes.
Stem cells translational medicine
Zhu Y,Jia Y,Wang Y,Xu J,Chai Y
Sat Jun 01 00:00:00 UTC 2019
Cholesterol Homeostatic Regulator SCAP-SREBP2 Integrates NLRP3 Inflammasome Activation and Cholesterol Biosynthetic Signaling in Macrophages.
Immunity
Guo C,Chi Z,Jiang D,Xu T,Yu W,Wang Z,Chen S,Zhang L,Liu Q,Guo X,Zhang X,Li W,Lu L,Wu Y,Song BL,Wang D
Tue Nov 20 00:00:00 UTC 2018
Cholesterol Homeostatic Regulator SCAP-SREBP2 Integrates NLRP3 Inflammasome Activation and Cholesterol Biosynthetic Signaling in Macrophages.
Immunity
Guo C,Chi Z,Jiang D,Xu T,Yu W,Wang Z,Chen S,Zhang L,Liu Q,Guo X,Zhang X,Li W,Lu L,Wu Y,Song BL,Wang D
Tue Nov 20 00:00:00 UTC 2018
A Golgi Lipid Signaling Pathway Controls Apical Golgi Distribution and Cell Polarity during Neurogenesis.
Developmental cell
Xie Z,Hur SK,Zhao L,Abrams CS,Bankaitis VA
Mon Mar 26 00:00:00 UTC 2018
A Golgi Lipid Signaling Pathway Controls Apical Golgi Distribution and Cell Polarity during Neurogenesis.
Developmental cell
Xie Z,Hur SK,Zhao L,Abrams CS,Bankaitis VA
Mon Mar 26 00:00:00 UTC 2018
Identification of MOSPD2, a novel scaffold for endoplasmic reticulum membrane contact sites.
EMBO reports
Di Mattia T,Wilhelm LP,Ikhlef S,Wendling C,Spehner D,Nominé Y,Giordano F,Mathelin C,Drin G,Tomasetto C,Alpy F
Sun Jul 01 00:00:00 UTC 2018
Identification of MOSPD2, a novel scaffold for endoplasmic reticulum membrane contact sites.
EMBO reports
Di Mattia T,Wilhelm LP,Ikhlef S,Wendling C,Spehner D,Nominé Y,Giordano F,Mathelin C,Drin G,Tomasetto C,Alpy F
Sun Jul 01 00:00:00 UTC 2018
Single-Cell Analysis of the Impact of Host Cell Heterogeneity on Infection with Foot-and-Mouth Disease Virus.
Journal of virology
Xin X,Wang H,Han L,Wang M,Fang H,Hao Y,Li J,Zhang H,Zheng C,Shen C
Tue May 01 00:00:00 UTC 2018
Single-Cell Analysis of the Impact of Host Cell Heterogeneity on Infection with Foot-and-Mouth Disease Virus.
Journal of virology
Xin X,Wang H,Han L,Wang M,Fang H,Hao Y,Li J,Zhang H,Zheng C,Shen C
Tue May 01 00:00:00 UTC 2018
Microglia Express Mu Opioid Receptor: Insights From Transcriptomics and Fluorescent Reporter Mice.
Frontiers in psychiatry
Maduna T,Audouard E,Dembélé D,Mouzaoui N,Reiss D,Massotte D,Gaveriaux-Ruff C
Wed Sep 30 00:00:00 UTC 2020
Microglia Express Mu Opioid Receptor: Insights From Transcriptomics and Fluorescent Reporter Mice.
Frontiers in psychiatry
Maduna T,Audouard E,Dembélé D,Mouzaoui N,Reiss D,Massotte D,Gaveriaux-Ruff C
Wed Sep 30 00:00:00 UTC 2020
Sam50 Regulates PINK1-Parkin-Mediated Mitophagy by Controlling PINK1 Stability and Mitochondrial Morphology.
Cell reports
Jian F,Chen D,Chen L,Yan C,Lu B,Zhu Y,Chen S,Shi A,Chan DC,Song Z
Tue Jun 05 00:00:00 UTC 2018
Sam50 Regulates PINK1-Parkin-Mediated Mitophagy by Controlling PINK1 Stability and Mitochondrial Morphology.
Cell reports
Jian F,Chen D,Chen L,Yan C,Lu B,Zhu Y,Chen S,Shi A,Chan DC,Song Z
Tue Jun 05 00:00:00 UTC 2018
Autophagy defects and related genetic variations in renal cell carcinoma with eosinophilic cytoplasmic inclusions.
Scientific reports
Yu Z,Ma J,Li X,Liu Y,Li M,Wang L,Zhao M,He H,Zhang Y,Rao Q,Zhao D,Wang Y,Fan L,Li P,Liu Y,Liu F,Zhang F,Ye J,Yan Q,Guo S,Wang Z
Mon Jul 02 00:00:00 UTC 2018
Autophagy defects and related genetic variations in renal cell carcinoma with eosinophilic cytoplasmic inclusions.
Scientific reports
Yu Z,Ma J,Li X,Liu Y,Li M,Wang L,Zhao M,He H,Zhang Y,Rao Q,Zhao D,Wang Y,Fan L,Li P,Liu Y,Liu F,Zhang F,Ye J,Yan Q,Guo S,Wang Z
Mon Jul 02 00:00:00 UTC 2018
Autophagy defects and related genetic variations in renal cell carcinoma with eosinophilic cytoplasmic inclusions.
Scientific reports
Yu Z,Ma J,Li X,Liu Y,Li M,Wang L,Zhao M,He H,Zhang Y,Rao Q,Zhao D,Wang Y,Fan L,Li P,Liu Y,Liu F,Zhang F,Ye J,Yan Q,Guo S,Wang Z
Mon Jul 02 00:00:00 UTC 2018
Protein kinase D at the Golgi controls NLRP3 inflammasome activation.
The Journal of experimental medicine
Zhang Z,Meszaros G,He WT,Xu Y,de Fatima Magliarelli H,Mailly L,Mihlan M,Liu Y,Puig Gámez M,Goginashvili A,Pasquier A,Bielska O,Neven B,Quartier P,Aebersold R,Baumert TF,Georgel P,Han J,Ricci R
Mon Sep 04 00:00:00 UTC 2017
Protein kinase D at the Golgi controls NLRP3 inflammasome activation.
The Journal of experimental medicine
Zhang Z,Meszaros G,He WT,Xu Y,de Fatima Magliarelli H,Mailly L,Mihlan M,Liu Y,Puig Gámez M,Goginashvili A,Pasquier A,Bielska O,Neven B,Quartier P,Aebersold R,Baumert TF,Georgel P,Han J,Ricci R
Mon Sep 04 00:00:00 UTC 2017
Protein kinase D at the Golgi controls NLRP3 inflammasome activation.
The Journal of experimental medicine
Zhang Z,Meszaros G,He WT,Xu Y,de Fatima Magliarelli H,Mailly L,Mihlan M,Liu Y,Puig Gámez M,Goginashvili A,Pasquier A,Bielska O,Neven B,Quartier P,Aebersold R,Baumert TF,Georgel P,Han J,Ricci R
Mon Sep 04 00:00:00 UTC 2017
Protein kinase D at the Golgi controls NLRP3 inflammasome activation.
The Journal of experimental medicine
Zhang Z,Meszaros G,He WT,Xu Y,de Fatima Magliarelli H,Mailly L,Mihlan M,Liu Y,Puig Gámez M,Goginashvili A,Pasquier A,Bielska O,Neven B,Quartier P,Aebersold R,Baumert TF,Georgel P,Han J,Ricci R
Mon Sep 04 00:00:00 UTC 2017
ESCRT-I Protein Tsg101 Plays a Role in the Post-macropinocytic Trafficking and Infection of Endothelial Cells by Kaposi's Sarcoma-Associated Herpesvirus.
PLoS pathogens
Kumar B,Dutta D,Iqbal J,Ansari MA,Roy A,Chikoti L,Pisano G,Veettil MV,Chandran B
Sat Oct 01 00:00:00 UTC 2016
ESCRT-I Protein Tsg101 Plays a Role in the Post-macropinocytic Trafficking and Infection of Endothelial Cells by Kaposi's Sarcoma-Associated Herpesvirus.
PLoS pathogens
Kumar B,Dutta D,Iqbal J,Ansari MA,Roy A,Chikoti L,Pisano G,Veettil MV,Chandran B
Sat Oct 01 00:00:00 UTC 2016
Quantitative proteomic analysis identifies new effectors of FOXM1 involved in breast cancer cell migration.
International journal of clinical and experimental pathology
Ye X,Zhang Y,He B,Meng Y,Li Y,Gao Y
Mon Oct 31 00:00:00 UTC 2016
Quantitative proteomic analysis identifies new effectors of FOXM1 involved in breast cancer cell migration.
International journal of clinical and experimental pathology
Ye X,Zhang Y,He B,Meng Y,Li Y,Gao Y
Mon Oct 31 00:00:00 UTC 2016
Characterization and proteomic analysis of ovarian cancer-derived exosomes.
Journal of proteomics
Liang B,Peng P,Chen S,Li L,Zhang M,Cao D,Yang J,Li H,Gui T,Li X,Shen K
Wed Mar 27 00:00:00 UTC 2013
Characterization and proteomic analysis of ovarian cancer-derived exosomes.
Journal of proteomics
Liang B,Peng P,Chen S,Li L,Zhang M,Cao D,Yang J,Li H,Gui T,Li X,Shen K
Wed Mar 27 00:00:00 UTC 2013
Therapeutic potential for phenytoin: targeting Na(v)1.5 sodium channels to reduce migration and invasion in metastatic breast cancer.
Breast cancer research and treatment
Yang M,Kozminski DJ,Wold LA,Modak R,Calhoun JD,Isom LL,Brackenbury WJ
Sun Jul 01 00:00:00 UTC 2012
Therapeutic potential for phenytoin: targeting Na(v)1.5 sodium channels to reduce migration and invasion in metastatic breast cancer.
Breast cancer research and treatment
Yang M,Kozminski DJ,Wold LA,Modak R,Calhoun JD,Isom LL,Brackenbury WJ
Sun Jul 01 00:00:00 UTC 2012
A novel non-canonical mechanism of regulation of MST3 (mammalian Sterile20-related kinase 3).
The Biochemical journal
Fuller SJ,McGuffin LJ,Marshall AK,Giraldo A,Pikkarainen S,Clerk A,Sugden PH
Thu Mar 15 00:00:00 UTC 2012