Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: LRAQLSLMAH PGEGDGLDRE EEEDEEEEEE EAVAVPQPMP SIPEDLESRE AMVAFFNSAV ASAEEEQARL RGQLKEQRVR CRRLAHLLAS AQKEPEAAAP APGTGGDSVC GETHRALQGA MEKLQSRFME LMQEKADLKE RVEELEHRCI QLSGETDTIG EYIALYQSQR AVLKERHREK EEYISRLAQD KEEMKVKLLE LQELVLRLVG DRNEWHGRFL AAAQNPADEP TSGAPAPQEL GAANQQGDLC EVSLAGSVEP AQGEAREGSP RDNPTAQQIM QLLREMQNPR ERPGLGSNPC IPFFYRADEN DEVKITVI
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130 kDa cis-Golgi matrix protein; cis-Golgi matrix protein GM130; GM130; GM130 autoantigen; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; Golgin subfamily A member 2; Golgin-95; RP11-395P17.5; SY11 protein
Gene Aliases: GM130; GOLGA2
UniProt ID: (Human) Q08379, (Rat) Q62839
Entrez Gene ID: (Human) 2801, (Rat) 64528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support