Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: STRGTYGILS KTVGTQDTPY MVNGQEIPAD TLGQFPSIKD VLQAAAAEHQ DQGQEVNGEV RSRRDSICSS VSLESSAAET QEEMLQVLKE KMRLEG
Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 71%.
The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCP170; golgi autoantigen, golgin subfamily a, 3; Golgi complex-associated protein of 170 kDa; Golgi membrane associated protein; Golgi peripheral membrane protein; Golgin subfamily A member 3; Golgin-160; golgin-165; HGNC:4426; male enhanced antigen-2; SY2/SY10 protein
Gene Aliases: GCP170; GOLGA3; MEA-2
UniProt ID: (Human) Q08378
Entrez Gene ID: (Human) 2802
Molecular Function: cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support