Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFKKLKQKIS EEQQQLQQAL APAQASSNSS TPTRMRSRTS SFTEQLDEGT PNRESGDTQS FAQKLQLRVP SVESLFRSPI KESLFRSSSK ESLVRTSSRE SLNRLDLDSS TASFDPPSDM DSEAEDLVGN SDSLNKEQLI QRLRRMERSL; Positive Samples: U-87MG, HeLa, SW480, A-549, MCF7, Mouse heart, Mouse brain; Cellular Location: Cytoplasm, Golgi apparatus, Golgi apparatus membrane, Peripheral membrane protein
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This protein has been postulated to play a role in Rab6-regulated membrane-tethering events in the Golgi apparatus. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 256 kDa golgin; 72.1 protein; centrosome-related protein F46; golga4 {ECO:0000312|EMBL:AAH85124.1, ECO:0000312|RGD:1591925}; golgi autoantigen, golgin subfamily a, 4; Golgin subfamily A member 4; golgin subfamily A member 4 {ECO:0000250|UniProtKB:Q13439}; golgin-240; Golgin-245; Protein 72.1; tGolgin-1; Trans-Golgi p230
Gene Aliases: AI225887; AU019508; CRPF46; GCP2; GOLG; GOLGA4; MU-RMS-40.18; Olp-1; p230
UniProt ID: (Human) Q13439, (Mouse) Q91VW5
Entrez Gene ID: (Human) 2803, (Mouse) 54214, (Rat) 501069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support