Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GOLGA7 may be involved in protein transport from Golgi to cell surface. The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: golgi autoantigen, golgin subfamily a, 7; Golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; Golgin subfamily A member 7
Gene Aliases: AB041568; C130038N16Rik; GCP16; GOLGA3AP1; GOLGA7; GOLGA7A; HDCKB03P; HSPC041; MNCb-1213; R75586
UniProt ID: (Human) Q7Z5G4, (Mouse) Q91W53, (Rat) Q6AYQ1
Entrez Gene ID: (Human) 51125, (Mouse) 57437, (Rat) 361171
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support