Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa heat shocked lysate.
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 27 kDa Golgi SNARE protein; Golgi SNAP receptor complex member 2; golgi SNARE; Membrin; RP23-272P17.5
Gene Aliases: 2310032N09Rik; Bos1; C76855; EPM6; GOSR2; GS27; I79_025823; membrin; SNARE
UniProt ID: (Human) O14653, (Rat) O35165, (Mouse) O35166
Entrez Gene ID: (Human) 9570, (Rabbit) 100342992, (Dog) 610436, (Pig) 100519332, (Rat) 64154, (Mouse) 56494, (Chinese hamster) 100766577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support