Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ22389; Nuclear envelope pore membrane protein POM 210; Nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; Nuclear pore protein gp210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; Nucleoporin Nup210; POM210; Pore membrane protein of 210 kDa
Gene Aliases: 9830001L10; AI836801; gp190; GP210; KIAA0906; NUP210; POM210; PSEC0245
UniProt ID: (Human) Q8TEM1, (Mouse) Q9QY81
Entrez Gene ID: (Human) 23225, (Mouse) 54563
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support