Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
GPR15 expression has been reported in CD4(+) T lymphocytes, alveolar macrophages, and the basal surface of the small intestinal epithelium. ESTs have been isolated from normal brain and kidney cancer libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BoB; Brother of Bonzo; G-protein coupled receptor 15; Gpr-15; MGC126828; MGC126830
Gene Aliases: 4933439K08Rik; BOB; GPR15
UniProt ID: (Human) P49685, (Mouse) Q0VDU3
Entrez Gene ID: (Human) 2838, (Mouse) 71223
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support