Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GPR75 is an Orphan-A GPCR with an unknown ligand. G protein-coupled receptor GPR75 has been reported to be expressed in brain and eye. ESTs have been isolated from human brain, eye, nerve, skin, and stomach libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor GPR75; GPR-chr2; Probable G-protein coupled receptor 75; WI-31133
Gene Aliases: GPR75; GPRchr2; WI31133
UniProt ID: (Human) O95800, (Mouse) Q6X632
Entrez Gene ID: (Human) 10936, (Mouse) 237716, (Rat) 498434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support