Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 95; G-protein coupled receptor 87; G-protein coupled receptor 95; MGC131898; orphan GPCR 87
Gene Aliases: FKSG78; GPR87; GPR95; KPG_002
UniProt ID: (Human) Q9BY21
Entrez Gene ID: (Human) 53836, (Bovine) 785030, (Rat) 310443
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support