Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
RXFP3 (Relaxin Family Peptide Receptor 3) is a Protein Coding gene. Diseases associated with RXFP3 include Schizotypal Personality Disorder and Chromosome 5P13 Duplication Syndrome. Among its related pathways are Signaling by GPCR and GPCRs, Other. Gene Ontology (GO) annotations related to this gene include G protein-coupled receptor activity and G protein-coupled peptide receptor activity. An important paralog of this gene is GPR132.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor SALPR; G protein-coupled receptor SALPR homolog; G-protein coupled receptor GPCR135; G-protein coupled receptor SALPR; MGC141998; MGC142000; relaxin 3 receptor 1; Relaxin family peptide receptor 3; Relaxin-3 receptor 1; relaxin-3/INSL7 receptor 1; relaxin/insulin-like family peptide receptor 3; RLN3 receptor 1; somatostatin and angiotensin-like peptide receptor; Somatostatin- and angiotensin-like peptide receptor
Gene Aliases: BC053073; GPCR135; RGD1311550; RLN3R1; RXFP3; RXFPR3; SALPR
UniProt ID: (Human) Q9NSD7, (Mouse) Q8BGE9
Entrez Gene ID: (Human) 51289, (Rat) 294807, (Mouse) 239336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support