Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MAALTRDPQ FQKLQQWYRE HRSELNLRRL FDANKDRFNH FSLTLNTNHG HILVDYSKNL VTEDVMRMLV DLAKSRGVEA ARERMFNGEK INYTEGRAVL HVALRNRSNT PILVDGKDVM PEVNKVLDKM KSFCQRVRSG DWKGYTGKTI TDVINIGIGG SDLGPLMVTE ALKPYSSGGP RVWYVSNIDG THIAKTLAQL NPESSLFIIA SKTFTTQETI TNAETAKEWF LQAAKDPSAV AKHFVALSTN TTKVKEFGID PQNMFEFWDW VGG (1-272 aa encoded by BC004982)
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMF; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; Glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250|UniProtKB:P06745}; GPI; GPI {ECO:0000250|UniProtKB:P06745}; gpi {ECO:0000312|RGD:2727}; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; Neuroleukin; neuroleukin {ECO:0000250|UniProtKB:P06745}; NLK; NLK {ECO:0000250|UniProtKB:P06745}; oxoisomerase; PGI; PGI {ECO:0000250|UniProtKB:P06745}; PHI; PHI {ECO:0000250|UniProtKB:P06745}; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250|UniProtKB:P06745}; phosphohexomutase; Phosphohexose isomerase; phosphohexose isomerase {ECO:0000250|UniProtKB:P06745}; phosphosaccharomutase; SA-36; sperm antigen; Sperm antigen 36; sperm antigen-36
Gene Aliases: AMF; GNPI; GPI; Gpi-1; Gpi-1r; Gpi-1s; Gpi-1t; Gpi1; Gpi1-r; Gpi1-s; Gpi1-t; Gpi1s; MF; NK; NK/GPI; NLK; Org; PGI; PHI; SA-36; SA36
UniProt ID: (Human) P06744, (Rat) Q6P6V0, (Mouse) P06745
Entrez Gene ID: (Human) 2821, (Rat) 292804, (Mouse) 14751
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support