Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Positive test controls include: Human plasma, Mouse plasma, Mouse liver, Rat plasma. The target is usually found in the following locations: Secreted.
Immunogen sequence: CGLSTHVEIG HRALEFLQLH NGRVNYRELL LEHQDAYQAG IVFPDCFYPS ICKGGKFHDV SESTHWTPFL NASVHYIREN YPLPWEKDTE KLVAFLFGIT SHMAADVSWH SLGLEQGFLR TMGAIDFHGS YSEAHSA
GPLD1 is expressed in numerous tissues and cells and specifically cleaves GPI-anchored proteins. Liver has the highest level of GPI-PLD expression and is the primary organ contributing to GPLD1 in the serum. GPLD1 is abundant in serum in which it associates with polipoproteins AI and AIV. Increased serum GPLD1 is associated with insulin resistance and elevated serum triglycerides. Many surface proteins are attached to eukaryotic cell membranes via glycosylphosphatidylinositol (GPI) anchors that are covalently bound to the C-terminus of the protein and cleavage of the GPI moiety by GPLD1, only enzyme known that cleavage GPI anchor, may represent a means of regulating attachment of these proteins to the cell surface, or alternatively, their release into the extracellular environment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycoprotein phospholipase D; Glycosyl-phosphatidylinositol-specific phospholipase D; glycosylphosphatidylinositol phospholipase D; glycosylphosphatidylinositol specific phospholipase D1, isoform 2; GPI-PLD; GPI-specific phospholipase D; MGC22590; Phosphatidylinositol-glycan-specific phospholipase D; PI-G PLD
Gene Aliases: 6330541J12Rik; AW546131; GPIPLD; GPIPLDM; GPLD1; PIGPLD; PIGPLD1; PLD
UniProt ID: (Human) P80108, (Rat) Q8R2H5
Entrez Gene ID: (Human) 2822, (Rat) 291132, (Mouse) 14756
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support